EDTA disodium salt dihydrate, Molecular Biology Grade, Calbiochem®

Supplier: MilliporeSigma

Synonyms: Ethylenediaminetetraacetic acid, disodium salt, dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium dihydrate, Titriplex III, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Sequestrene Na₂, Edetate disodium dihydrate, Edetic acid disodium salt dihydrate

324503-100GM 324503-1KG
80502-380EA 85.32 USD
80502-380 80502-382
EDTA disodium salt dihydrate, Molecular Biology Grade, Calbiochem®
EDTA disodium salt dihydrate
Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24 g/mol
Melting Pt: 255
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White crystalline solid
Contaminants DNases, proteases, RNases: none detected
Storage Store at 15 to 30 °C, Hygroscopic
Stability Following reconstitution, store at room temperature (20 °C). Stock solutions are stable for up to 6 months at 20 °C.
Solubility H₂O (186 mg/mL or 0.5M) Adjust pH to 8.0 for complete solubilization.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR