Dihydroethidium ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 3,8-Diamino-5,6-dihydro-5-ethyl-6-phenylphenanthridine, Hydroethidine, DHE, 5-Ethyl-6-phenyl-5,6-dihydrophenanthridine-3,8-diamine, 2,7-Diamino-10-ethyl-9-phenyl-9,10-dihydrophenanthridine

309800-25MG
80502-356EA 288.38 USD
80502-356
Dihydroethidium ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
Dihydroethidium

A cell-permeable, chemically-reduced ethidium derivative.

Formula: C₂₁H₂₁N₃
Storage Temperature: Freezer
MDL Number: MFCD00077335
CAS Number: 104821-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR