Calcium chloride dihydrate ≥99% (by complexometric analysis), white solid, Molecular Biology Grade for molecular biology, Calbiochem®

Supplier: MilliporeSigma
Warning

208291-250GM
80501-994EA 62.55 USD
80501-994
Calcium chloride dihydrate ≥99% (by complexometric analysis), white solid, Molecular Biology Grade for molecular biology, Calbiochem®
Calcium chloride dihydrate
Formula: CaCl₂·2H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149613
CAS Number: 10035-04-8

Order Now

Specification Test Results

Purity (by complexometry) ≥99%
Heavy Metals ≤0.0005% (as Pb)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR