2-Mercaptoethanol ≥99%, Molecular Biology Grade, Calbiochem®

Supplier: MilliporeSigma

Synonyms: beta-Mercaptoethanol, 2-Hydroxyethyl mercaptan, β-Mercaptoethanol

444203-250ML
80501-154EA 128.27 USD
80501-154
2-Mercaptoethanol ≥99%, Molecular Biology Grade, Calbiochem®
2-Mercaptoethanol

β-Mercaptoethanol, Molecular Biology Grade a disulfide-reducing agent.

Formula: HSCH₂CH₂OH
MW: 78.14 g/mol
MDL Number: MFCD00004890
CAS Number: 60-24-2
UN: 2966
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc427209b84d0343a5ae47c435ddfc53df4013d2&vid=7d70fd087d948d54e7a946d3b2310e2d820d2396&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥99%
DNases, proteases, RNases none detected.
Absorbance (260 nm) ≤1.50
Absorbance (280 nm) ≤0.31

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR