Phenol, Molecular Biology Grade

Supplier: MilliporeSigma

Synonyms: Phenylalcohol, Carbolic acid, Hydroxybenzene

516724-500GM
80501-126EA 278.19 USD
80501-126
Phenol, Molecular Biology Grade
Phenol
Formula: C₆H₅OH
MW: 94.11 g/mol
Boiling Pt: 181.7
Melting Pt: 40.8
Flash Pt: 81
MDL Number: MFCD00002143
CAS Number: 108-95-2
Merck Index: 12,07390

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR