Dansyl chloride ≥99% (by HPLC), Calbiochem®, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

311155-1GM
80501-034EA 95.64 USD
80501-034
Dansyl chloride ≥99% (by HPLC), Calbiochem®, Sigma-Aldrich®
Dansyl chloride

Reagent used for fluorescent labeling of proteins, N-terminal amino acids, and amines.

Formula: C₁₂H₁₂ClNO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD00003985
CAS Number: 605-65-2
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR