NovaTaq™ PCR Master Mix Kit, Novagen®, MilliporeSigma
Supplier: MilliporeSigma
71007-3
80500-368EA 260.68 USD
80500-368
NovaTaq™ PCR Master Mix Kit, Novagen®, MilliporeSigma
Nucleic Acid Reagents End-point PCR Enzymes and Kits
The NovaTaq™ PCR Master Mix is a ready-to-use mixture of ultrapure deoxynucleotides
Offers advantages of time-savings, consistency, and minimal risk of contamination. Supplied in package sizes of 100rxn (100µL/rxn).
Components: 2X concentrated NovaTaq DNA polymerase, Ultrapure deoxynucleotides, Reaction buffer without MgCl2, 4x1.25 mL for 2x Nova Taq PCR Master Mix, 1.5 mL for 25 mM MgCl2 solution, 6 mL for PCR grade water.