NovaTaq™ DNA Polymerase, Novagen®, MilliporeSigma
Supplier: MilliporeSigma
71003-5 71003-4
80500-362EA 1210.93 USD
80500-362 80500-360
NovaTaq™ DNA Polymerase, Novagen®, MilliporeSigma
Nucleic Acid Reagents End-point PCR Enzymes and Kits
This polymerase is a 94 kDa premium-quality recombinant form of Taq DNA polymerase.
This thermostable enzyme is suitable for a wide range of PCR applications. To ensure the highest purity and reproducible performance, each preparation is extensively tested in a variety of quality control assays. NovaTaq DNA Polymerase possesses an integral 5'–3' exonuclease and lacks a 3'–5' exonuclease activity. The enzyme leaves single 3' nucleotide overhangs that make the products suitable for cloning by T–vector or U–vector methods.
Ordering information: Each order also includes optimized 10X PCR buffers (750 µl), with and without MgCl₂, and 25 mM MgCl₂ solution (1.5 ml).
Packaging: Packed in glass bottles.