Calbiochem® Ubiquitin Aldehyde
Supplier: MilliporeSigma
662056-50UG
80500-114EA 354.47 USD
80500-114
Calbiochem® Ubiquitin Aldehyde
Proteins and Peptides
Potent and specific inhibitor of multiple ubiquitin hydrolases involved in pathways of intracellular protein modification and turnover
Bioassay: shown to inhibit the activity of isopeptidase T (80052-740) and ubiquitin C-terminal hydrolase (80054-944).
Useful for stabilizing endogenous or in vitro synthesized ubiquitin-protein conjugates. May also be used to enhance or decrease rates of ubiquitin-dependent degradation.