[beta]-Amyloid Peptide (1-42)
Supplier: MilliporeSigma
PP69-.25MG
80500-030EA 490.14 USD
80500-030
[beta]-Amyloid Peptide (1-42)
Proteins and Peptides
Synthetic peptide corresponding to amino acids 1–42 of the processed human amyloid peptide. Supplied in a form that is not neurotoxic prior to preincubation. The level of toxicity has recently been shown to correlate to the extent of beta sheet structure. Reconstitute with degassed HPLC grade deionized water.