β-Carotene ≥97%, Calbiochem®

Supplier: MilliporeSigma

Synonyms: 2,2'-((1E,3E,5E,7E,9E,11E,13E,15E,17E)-3,7,12,16-Tetramethyloctadeca-1,3,5,7,9,11,13,15,17-nonaene-1,18-diyl)bis(1,3,3-trimethylcyclohex-1-ene), Provitamin A

Packaged under inert gas.

217538-1GM
80109-972EA 121.94 USD
80109-972
β-Carotene ≥97%, Calbiochem®
beta-Carotene

β-Carotene, Vitamin A precursor.


  • Dark red solid form
  • Soluble in DMSO or cyclohexane

Formula: C₄₀H₅₆
MW: 536.89 g/mol
MDL Number: MFCD00001556
CAS Number: 7235-40-7

Order Now

Specification Test Results

Purity by UV ≥97%
Storage -20°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR