Tocopherol Set, Calbiochem®, EMD Millipore
Supplier: MilliporeSigma
613424-1SET
80109-952EA 1576.71 USD
80109-952
Tocopherol Set, Calbiochem®, EMD Millipore
Standards Organic Standards
The Tocopherol set is to be used as a quantitative standard, and the material should be weighed analytically before use
  • ≥95% by HPLC (for each component)
  • Packaged under inert gas
If small quantities of tocopherol are to be used at one time, slowly draw the material from the vial using a Hamilton syringe, flush with inert gas, and re-seal the vial.
Delivery information: Tocopherol Set is a convenient four-vial pack containing ~50 mg each of all-rac-α, all-rac-β, RRR-λ, and RRR-δ-tocopherols.