Riboflavine (vitamin B2) 99.0-101.0%, Calbiochem®

Supplier: MilliporeSigma

Synonyms: Vitamin B2, Lactoflavin, 7,8-Dimethyl-10-((2S,3S,4R)-2,3,4,5-tetrahydroxypentyl)benzo[g]pteridine-2,4(3H,10H)-dione, (-)-Riboflavin, Vitamine G

555682-10GM
80109-776EA 71.44 USD
80109-776
Riboflavine (vitamin B2) 99.0-101.0%, Calbiochem®
Riboflavine (Vitamin B2)

A water soluble vitamin involved in the synthesis of flavin mononucleotide (FMN) and flavin adenine dinucleotide (FAD). Provides protection against oxidative damage.

Formula: C₁₇H₂₀N₄O₆
MW: 376.37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005022
CAS Number: 83-88-5

Order Now

Specification Test Results

Synonyms Vitamin B₂
Form Yellow-Orange Solid
Hill Formula C17H20N4O6
Chemical Formula C17H20N4O6
Optical Rotation -135.0° to -155.0° (c = 0.5%, 50 mM NaOH, dry basis)
Purity 99.0 - 101.0% by UV
Contaminants Conforms (Lumiflavine)
Toxicity Standard Handling
Storage 15 to 30 °C
Protect from Light Protect from Light
Do not Freeze Ok to Freeze
Special Instructions Following reconstitution, aliquot and freeze (-20 °C). Stock solutions are stable for up to 3 months at -20 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR