L(-)-Sorbose

Supplier: MilliporeSigma

Synonyms: L-(-)-Sorbose, (3S,4R,5S)-1,3,4,5,6-Pentahydroxyhexan-2-one

567692-1KG 567692-250GM
80109-422EA 1905.36 USD
80109-422 80109-424
L(-)-Sorbose
L(-)-Sorbose
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 380
MDL Number: MFCD00151097
CAS Number: 87-79-6
Merck Index: 12,08874

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR