Human Recombinant Caspase-1 (from E. coli)
Supplier: MilliporeSigma
218783-3000U
80108-900EA 608.27 USD
80108-900
Human Recombinant Caspase-1 (from E. coli)
Proteins and Peptides
In 500mM NaCl, 150mM imidazole, 20mM Tris-HCl, 5mM DTT, 1mM EDTA, 10% glycerol, 0.1% CHAPS, pH 7.9
Useful for the study of enzyme regulation, cleavage of target substrates, and inhibitor screening. Activity: >=50,000units/mL. Specific activity: >=25,000units/mg protein.