Uracil ≥99% (by HPLC), Calbiochem®, Millipore®

Supplier: MilliporeSigma

Synonyms: Pyrimidine-2,4(1H,3H)-dione

6630-10GM
80108-324EA 71.63 USD
80108-324
Uracil ≥99% (by HPLC), Calbiochem®, Millipore®
Uracil
Formula: C₄H₄N₂O₂
MW: 112.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006016
CAS Number: 66-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR