Nicotinic acid (Niacin)

Supplier: MilliporeSigma

Synonyms: 3-Pyridinecarboxylic acid, Pyridine-3-carboxylic acid, 3-Picolinic acid, Niacin, Nicotinic acid, Vitamin B3

481918-100GM
80108-214EA 83.85 USD
80108-214
Nicotinic acid (Niacin)
Nicotinic acid (Niacin)
Formula: C₆H₅NO₂
MW: 123.11 g/mol
MDL Number: MFCD00006391
CAS Number: 59-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR