L(+)-Cysteine free base ≥98% (by titrimetric analysis), Calbiochem®, Millipore®

Supplier: MilliporeSigma

Synonyms: H-Cys-OH, L-(+)-Cysteine, (R)-2-Amino-3-mercaptopropanoic acid

Packaged under inert gas.

243005-1KG 243005-25GM
80108-178EA 382.07 USD
80108-178 80108-180 80108-182
L(+)-Cysteine free base ≥98% (by titrimetric analysis), Calbiochem®, Millipore®
L-Cysteine

Naturally-occurring non-essential amino acid.

Formula: HSCH₂CH(NH₂)CO₂H
MW: 121.16 g/mol
Boiling Pt: 254
Melting Pt: 115
Storage Temperature: Ambient
MDL Number: MFCD00064306
CAS Number: 52-90-4
Merck Index: 14,02781

Order Now

Specification Test Results

Purity (by Titration, Perchloric Acid) ≥98%
Iron ≤10 ppm
Endotoxin ≤24 EU/g
Total microorganisms ≤100 CFU/g
Heavy metals ≤0.001% (as Pb)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR