Cladribine ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: (2R,3S,5R)-5-(6-Amino-2-chloro-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol, CdA, 2-Chloro-2'-deoxyadenosine, NSC105014, RWJ 26251

220467-10MG
80108-008EA 85.9 USD
80108-008
Cladribine ≥95% (by HPLC), Calbiochem®, Sigma-Aldrich®
Cladribine

A deoxyadenosine analog that displays strong antileukemic and immunosuppressive activity


Also induces apoptosis in chronic lymphocytic leukemia (CLL) cells.

Formula: C₁₀H₁₂ClN₅O₃
MW: 285.7 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00153939
CAS Number: 4291-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR