Hand-Held Alcohol Refractometers, AL-21[alpha], ATAGO®
Supplier: Atago
2361
80094-056EA 1012.62 USD
80094-056
Hand-Held Alcohol Refractometers, AL-21[alpha], ATAGO®
Refractometers Handheld Refractometers
Model AL-21[alpha] Refractometers is ideal for measuring pure and distilled alcohol
Ordering information: A calibration certificate is available. Contact your Avantor representative for information.