ActivArmr® 78-150 Lightweight Polyester Gloves, Ansell
Supplier: Ansell Healthcare
222005
80092-716CS 1297.71 USD
80092-716
ActivArmr® 78-150 Lightweight Polyester Gloves, Ansell
Gloves
Use as a hand warmer in cold, damp working environments or as an under glove.
  • Thermal insulation for cold weather protection
  • Automatic knit technology for long more comfortable wear
  • Can be worn beneath other gloves
  • Ambidextrous
  • Constructed of lightweight, warming materials
May be worn on either hand.
Use in the construction, food handling, catering, and public utilities industries. Uses include handling chilled or refrigerated products; fish processing; construction work; general maintenance; utilities; and general outdoor work.
Constructed of Thermolite® fabric and polyester. Lightweight.
Certifications: ANSI level 1 cut protection. ANSI level 3 abrasion protection.
Packaging: Gloves are packaged 12 pairs in a bag; 144 in a carton.