ActivArmr® 23-191 Cold resistant gloves, Ansell
Supplier: Ansell Healthcare
204968
80092-580CS 1846.54 USD
80092-580
ActivArmr® 23-191 Cold resistant gloves, Ansell
Gloves
These vinyl-dipped gloves with knit wrists are useful for keeping hands warm and protected from minor cuts and abrasions.
  • Vinyl-dipped coating on a thermal liner
  • Keeps hands warm, dry and protected from abrasions
  • PVC coating provides oil and grease repellence and remains flexible at low temperatures
Use in the general carpentry, janitorial services, and warehousing industries. Uses include cold storage and unheated warehouses; handling chilled or refrigerated products; outdoor construction work; patrolling unheated premises; refuse collection; and handling cartons and packaging during shipping and receiving.
Certifications: ANSI level 1 cut protection.
Packaging: Gloves are packaged 12 pairs in a bag; 6 bags in a carton.