Heat-Resistant Terrycloth Gloves, Wells Lamont
Supplier: Wells Lamont
1966
80091-274EA 9.6 USD
80091-274
Heat-Resistant Terrycloth Gloves, Wells Lamont
Gloves
Heavyweight, seamless gloves feature superior cut and abrasion protection.
  • Seamless
  • White
An insulating layer of air forms beneath each loop of fabric, allowing the gloves to dissipate heat quickly.
The ambidextrous, knit-wrist gloves protect against up to 176 °C (350 °F).