HEATBLOK Double Layer Palm Terry Cloth Gloves, Loop In, Wells Lamont
Supplier: Wells Lamont
320
80091-230EA 19.12 USD
80091-230
HEATBLOK Double Layer Palm Terry Cloth Gloves, Loop In, Wells Lamont
Gloves
These gloves resist heat up to 460 °F.
  • Safety cuff
  • Clute cut
Users can clean the ambidextrous white gloves in a washing machine.
Ordering information: Gloves are sold by the dozen pairs.
Caution: Caution Information: The degree of heat resistance a glove may offer is directly related to the duration of exposure and weight of the object being handled.