Cryogenic Printable Labels, Brady
Supplier: Brady Worldwide
116415 116412 116392 116409 116388 116407 116413 142210 142211 142212 66267 66265 66268 66271 66273 66274 105124 62357 62376 62377 62378 11265 11267 62355 62363 62366 62367 62369 30113 30221 35244 66270 66272 108241 11266 62372 62373 30176 62361 30127 20822 110382 62368 62370 11263 11264 62365 62371 62379 66269 66266 19260 108240 62375 30543 62354 62359 62364 62374 19261 35246 143119 110380 IP-R6406 110381 11510 10615 35269 54802 66263 66261 THT-122-461-3 THT-123-461-3 THT-126-499-3 THT-127-461-3 THT-131-427-3 THT-133-461-3 THT-136-499-3 THT-137-499-3 THT-139-461-2 THT-141-488-3 THT-151-499-3 THT-152-492-3 THT-152-499-3 THT-15-423-2.5 29895 THT-155-490-3 THT-156-490-3 THT-157-490-3 THT-163-499-3 THT-164-488-1 THT-179-492-3 THT-194-481-3 30513 THT-144-492-3 20838 30132 30135 30189 30207 134098 134097 30190 30188 104788 104789 104790
89175-190RL 251.42 USD
89175-190 89175-192 89175-198 89175-218 89175-230 89175-234 89175-236 89175-256 89175-258 89175-260 89009-232 89077-354 89077-360 89077-366 89077-370 89077-372 89085-400 80090-224 80090-234 80090-236 80090-238 80090-246 80090-250 80090-258 80090-262 80090-266 80090-278 80090-280 80090-282 80090-286 89006-918 89077-364 89077-368 89209-554 89209-558 94024-530 80090-248 80090-252 80090-254 80090-272 80090-274 80090-284 80090-290 89091-810 80090-228 80090-230 80090-242 80090-244 80090-264 80090-268 80090-270 89009-234 89077-356 89091-808 89209-486 94024-528 80090-232 80090-240 80090-256 80090-260 80090-276 80090-294 94024-532 94024-534 89066-904 53520-198 490006-114 490006-116 490006-118 490006-268 490006-270 490006-274 490006-276 490006-284 490006-290 490006-292 490006-294 490006-296 490006-298 490006-300 490006-302 490006-304 490006-306 490006-308 490006-310 490006-312 490006-314 490006-316 490006-318 490006-322 490006-326 490006-328 490006-330 490006-332 490006-334 490006-338 490006-340 490006-342 490006-346 490006-348 490006-350 490006-354 490006-356 490006-358 490006-362 490006-368 490006-376 490006-378 490006-380 490014-320 490016-396 10796-208 10796-210 10796-212 10796-214 10796-216 10796-488 10796-490 10796-506 10796-508 89209-491 89209-493 89209-495
Cryogenic Printable Labels, Brady
Labels
Cryogenic labels for tubes, vials and canes.
Chemical and stain resistant labels for slides and cassettes:
Temperatures from –80 to 121 °C (–112 to 250 °F).
B-449 Slide label with removability and write on capabilities.
B-481 Stainerbondz™ extreme chemical resistant slide label.
B-482 Tissue cassette label applied pre-process with the BSP®31 for extreme solvent/stain resistance.
B-488 Slide label with moderate chemical.
B-498 Tissue cassette label for re-identification of processed paraffin blocks.
General labels for straws, petri dishes, bottles, flasks, microplates, bags, and more:
B-424 Economical paper label for general purpose.
B-427 Straw label for wrapping; working temperatures from –196 to 80 °C (–321 to 176 °F).
B-430 Petri dish and bag label; working temperatures from –80 to 121 °C (–112 to 250 °F).
B-499 For cryo boxes, freezer racks, petri dishes, bottles, flasks, microplates, and bags; working temperatures from –196 to 121 °C (–321 to 250 °F).