PC Link Labels, Brady
Supplier: Brady Worldwide
18705
80090-202EA 156.62 USD
80090-202 80090-200 80090-208 80090-148 490006-206 490006-208 490006-210 490006-212 490006-214 490006-216 490006-218 490006-220 490006-222 490006-224 490006-226 490006-228 490006-230 490006-232 490006-234 490006-236 490006-238 490006-240 490006-242 490006-244 490006-246 490006-248 490006-250 490006-252 490006-254 490006-256 490006-258 490006-260 490006-262 490006-264 490006-266 490006-272 490006-280 490006-286
PC Link Labels, Brady
Labels
B-461: Self-laminating polyester is a clear film with a matte white printable zone ideal for cables, laboratory ID or general labeling where graphics need to be protected.
B-490: Durable polyester markers are designed for use in laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws and slides.
B-499: Nylon cloth wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable.
B-423: Permanent polyester is ideal for electronic PCB and component identification, bar code labels, rating plates and solar panel identification.
B-427: Self-laminating vinyl is an excellent material for wire and cable identification. This material has good clarity, conformability and is self-extinguishing.
B-351: Tamper-resistant vinyl is designed to fracture easily to show signs of product tampering and to prevent on-piece label removal.
B-435: Glossy metallized permanent polyester is designed for high adhesion to textured metals, powder coated surfaces and low surface energy plastics.
B-488: High performance polyester withstands numerous solvents and variable temperatures when applied to various surfaces including stainless steel and polypropylene.