BD Vacutainer® Push Button Blood Collection Sets, BD Diagnostics
Supplier: BD
367341 367342 367324 367344 368656 367324
95039-196CS 891.78 USD
95039-196 95030-190 89005-124 89005-126 80090-066 80090-068 95030-188 BD367344 BD367352 BD368656 BD368659 75795-904
BD Vacutainer® Push Button Blood Collection Sets, BD Diagnostics
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Blood collection sets are intended for use in blood collection and short-term infusion.
The in-vein needle retraction is comfortable for the patient and reduces the risk of exposure to contaminated needles for the healthcare worker.The push button feature provides immediate protection against injury or contamination
One-handed activation of the safety mechanism also allows the healthcare worker to attend to the patient and puncture site safely. The translucent body and tubing show blood flashback clearly. Blood collection sets are supplied sterile, and are latex- and DEHP-free.
Ease of use reduces training requirements and improves activation compliance.