Sodium chloride solution 0,9% for injection

Supplier: BAXTER IV SOLUTIONS

Synonyms: Saline, NaCl, Sea salt, Halite

2B1324X 2B1324X
80089-766CS 548.26 USD
80089-766 SC80089-766
Sodium chloride solution 0,9% for injection
Sodium chloride

  • Specific gravity: 1.01
  • Fill range volume (ml): 1030 to 1070
  • Store at room temperature (25 °C)
  • mOsmol/L (calc): 308
  • NDC Number: 0338-0049-04
  • pH: 5.0


Certifications: USP specified.

Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Merck Index: 14,08599

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR