Water USP for injection (WFI), sterile

Supplier: BAXTER IV SOLUTIONS

Synonyms: Dihydrogen oxide

2B0304X
80089-742CS 1107.56 USD
80089-742
Water USP for injection (WFI), sterile
Water

Sterile water for injection. For drug diluent use.


  • Specific gravity: 1
  • Fill range volume (ml): 1030 to 1070
  • Store at room temperature (25 °C)
  • pH 5.5
  • NDC number: 0338-0013-04

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR