Whatman™ Aqueous IFD, Whatman products (Cytiva)
Supplier: Cytiva
6726-5002 6726-5002A
28137-970PK 262.2 USD
28137-970 80089-524
Whatman™ Aqueous IFD, Whatman products (Cytiva)
Filters In-Line Filters
Aqueous IFD from Cytiva is a polypropylene housed in-line filter/degasser (IFD) with nylon membrane. It connects to an HPLC line and simultaneously filters and degasses during HPLC mobile phase preparation.
  • Faster than traditional methods of HPLC mobile phase preparation, saving time in the laboratory
  • Provides pure filtration of aqueous mobile phases without expensive degassing equipment
  • Chemically resistant polypropylene construction enables reliable filtration
  • Can be integrity-tested with the bubble point method
  • Includes compression end fittings
More Product Information