Anti-TPIPA Rabbit Polyclonal Antibody

Supplier: Abgent
AP6810A
80089-220EA 763.01 USD
80089-220
Anti-TPIPA Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TPIPA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR