Anti-DSS1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP6264A
80089-170EA 763.01 USD
80089-170
Anti-DSS1 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-DSS1 (N-term) Reacts with Human

Type: Primary
Antigen: DSS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR