Anti-SCDGFB Rabbit Polyclonal Antibody

Supplier: Abgent
AP6263A
80089-168EA 801.16 USD
80089-168
Anti-SCDGFB Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SCDGFB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR