Anti-GPRC5A Rabbit Polyclonal Antibody

Supplier: Abgent
AP6239A
80089-122EA 801.16 USD
80089-122
Anti-GPRC5A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GPRC5A (G protein-coupled receptor, class C, group 5, member A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR