Anti-FAM160B2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP6235A
80089-114EA 763.01 USD
80089-114
Anti-FAM160B2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FAM160B2 (family with sequence similarity 160, member B2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR