Anti-NOTCH1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP6218A
80089-080EA 801.6 USD
80089-080
Anti-NOTCH1 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-NOTCH1 (C-term) Reacts with Human

Type: Primary
Antigen: NOTCH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR