Anti-MMP28 Rabbit Polyclonal Antibody

Supplier: Abgent
AP6209A
80089-062EA 763.01 USD
80089-062
Anti-MMP28 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MMP28
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR