Anti-MLL2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP6183A
80089-010EA 808.65 USD
80089-010
Anti-MLL2 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-MLL2 (C-term) Reacts with Human

Type: Primary
Antigen: MLL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (4745–4759 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR