Anti-MAGEA3 Rabbit Polyclonal Antibody

Supplier: Abgent
AP6165A
80088-974EA 801.6 USD
80088-974
Anti-MAGEA3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MAGEA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR