Anti-LRP6 Rabbit Polyclonal Antibody

Supplier: Abgent
AP6158A
80088-960EA 801.36 USD
80088-960
Anti-LRP6 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LRP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (1500–1575 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR