Anti-LRP4 Rabbit Polyclonal Antibody

Supplier: Abgent
AP6156A
80088-956EA 763.01 USD
80088-956
Anti-LRP4 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-LRP4 (C-term) Reacts with Human

Type: Primary
Antigen: LRP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (1375–1425 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR