Anti-LRP2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP6154A
80088-952EA 801.16 USD
80088-952
Anti-LRP2 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-LRP2 (Megalin) (C-term) Reacts with Human

Type: Primary
Antigen: LRP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (4600–4675 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR