Anti-LEPR Rabbit Polyclonal Antibody

Supplier: Abgent
AP6151A
80088-946EA 811.49 USD
80088-946
Anti-LEPR Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LEPR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal (1-60 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR