Anti-XIAP Rabbit Polyclonal Antibody

Supplier: Abgent
AP6125A
80088-894EA 801.16 USD
80088-894
Anti-XIAP Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: XIAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal (1–50 aa)
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR