Anti-BIRC1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP6123A
80088-890EA 762.8 USD
80088-890
Anti-BIRC1 Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-BIRC1 (C-term) Reacts with Human

Type: Primary
Antigen: BIRC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR