Anti-ABCB7 Rabbit Polyclonal Antibody

Supplier: Abgent
AP6114A
80088-874EA 800.94 USD
80088-874
Anti-ABCB7 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ABCB7 (ATP-binding cassette, sub-family B (MDR/TAP), member 7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR