Anti-APBB2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP6101A
80088-850EA 763.01 USD
80088-850
Anti-APBB2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: APBB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR