Anti-SARS virus Spike Rabbit Polyclonal Antibody

Supplier: Abgent
AP6009C
80088-822EA 763.01 USD
80088-822
Anti-SARS virus Spike Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-SARS virus Spike (Center K439) Reacts with SARS Virus

Type: Primary
Antigen: SARS virus Spike (Center K439)
Clonality: Polyclonal
Clone:
Conjugation: (none)
Epitope:
Host: Rabbit
Isotype:
Reactivity: SARS Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR