Anti-SAE1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP2511B
80088-700EA 801.16 USD
80088-700
Anti-SAE1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SAE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR