Anti-PML Rabbit Polyclonal Antibody

Supplier: Abgent
AP2504B
80088-680EA 801.16 USD
80088-680
Anti-PML Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PML
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: 484–499
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR